missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Minpp1 Protein
A DNA sequence encoding the Homo sapiens (Human) Minpp1, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP8249-ec-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_001171588.1 | |
Minpp1 Protein | |
Research Use Only | |
Minpp1 | |
Recombinant Protein | |
E. coli | |
SLLEPRDPVA SSLSPYFGTK TRYEDVNPVL LSGPEAPWRD PELLEGTCTP VQLVALIRHG TRYPTVKQIR KLRQLHGLLQ ARGSRDGGAS STGSRDLGAA LADWPLWYAD WMDGQLVEKG RQDMRQLALR LASLFPALFS RENYGRLRLI TSSKHRCMDS SAAFLQGLWQ HYHPGLPPPD VADMEFGPPT VNDKLMRFFD HCEKFLTEVE KNATALYHVE AFKTGPEMQN ILKKVAATLQ VPVNDLNADL IQVAFFTCSF DLAIKGVKSP WCDVFDIDDA KVLEYLNDLK QYWKRGYGYT INSRSSCTLF QDIFQHLDKA VEQKQRSQPI SSPVILQFGH AETLLPLLSL MGYFKDKEPL TAYNYKKQMH RKFRSGLIVP YASNLIFVLY HCENAKTPKE QFRVQMLLNE KVLPLAYSQE TVSFYEDLKN HYKDILQSCQ TSEECELARA NSTSDEL | |
Greater than 90% as determined by SDS-PAGE. |
9562 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |