missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mitogen-activated protein kinase 6 Protein
A DNA sequence encoding the Homo sapiens (Human) Mitogen-activated protein kinase 6, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6341-ec-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_002739.1 | |
Mitogen-activated protein kinase 6 Protein | |
Research Use Only | |
MAPK6 | |
Recombinant Protein | |
E. coli | |
MAEKFESLMN IHGFDLGSRY MDLKPLGCGG NGLVFSAVDN DCDKRVAIKK IVLTDPQSVK HALREIKIIR RLDHDNIVKV FEILGPSGSQ LTDDVGSLTE LNSVYIVQEY METDLANVLE QGPLLEEHAR LFMYQLLRGL KYIHSANVLH RDLKPANLFI NTEDLVLKIG DFGLARIMDP HYSHKGHLSE GLVTKWYRSP RLLLSPNNYT KAIDMWAAGC IFAEMLTGKT LFAGAHELEQ MQLILESIPV VHEEDRQELL SVIPVYIRND MTEPHKPLTQ LLPGISREAL DFLEQILTFS PMDRLTAEEA LSHPYMSIYS FPMDEPISSH PFHIEDEVDD ILLMDETHSH IYNWERYHDC QFSEHDWPVH NNFDIDEVQL DPRALSDVTD EEEVQVDPRK YLDGDREKYL EDPAFDTNYS TEPCWQYSDH HENKYCDLEC SHTCNYKTRS SSYLDNLVWR ESEVNHYYEP KLIIDLSNWK EQSKEKSDKK GKSKCERNGL VKAQIALEEA SQQLAGKERE KNQGFDFDSF IAGTIQLSSQ HEPTDVVDKL NDLNSSVSQL ELKSLISKSV SQEKQEKGMA NLAQLEALYQ SSWDSQFVSG GEDCFFINQF CEVRKDEQVE KENTYTSYLD KFFSRKEDTE MLETEPVEDG KLGERGHEEG FLNNSGEFLF NKQLESIGIP QFHSPVGSPL KSIQATLTPS AMKSSPQIPH QTYSSILKHL N | |
Greater than 90% as determined by SDS-PAGE. |
5597 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |