missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Myosin regulatory lc 2, ventric/card muscle isoform Protein

A DNA sequence encoding the Homo sapiens (Human) Myosin regulatory lc 2, ventricular/cardiac muscle isoform, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP8680-ec-100ug

Weitere Details : net-weight : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16085921

  • 530.00 € / 100 Mikrogramm
Estimated Shipment: 07-02-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Beschreibung

Beschreibung

Spezifikation

Spezifikation

NP_000423.2
Myosin regulatory lc 2, ventric/card muscle isoform Protein
Research Use Only
MYL2
Recombinant Protein
E. coli
KRAGGANSNV FSMFEQTQIQ EFKEAFTIMD QNRDGFIDKN DLRDTFAALG RVNVKNEEID EMIKEAPGPI NFTVFLTMFG EKLKGADPEE TILNAFKVFD PEGKGVLKAD YVREMLTTQA ERFSKEEVDQ MFAAFPPDVT GNLDYKNLVH IITHGEE
Greater than 90% as determined by SDS-PAGE.
4633
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote