missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NAE1 Protein
A cDNA sequence encoding the NAE1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12783-10ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NAE1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAQLGKL LKEQKYDRQL RLWGDHGQEA LESAHVCLIN ATATGTEILK NLVLPGIGSF TIIDGNQVSG EDAGNNFFLQ RSSIGKNRAE AAMEFLQELN SDVSGSFVEE SPENLLDNDP SFFCRFTVVV ATQLPESTSL RLADVLWNSQ IPLLICRTYG LVGYMRIIIK EHPVIESHPD NALEDLRLDK PFPELREHFQ SYDLDHMEKK DHSHTPWIVI IAKYLAQWYS ETNGRIPKTY KEKEDFRDLI RQGILKNENG APEDEENFEE AIKNVNTALN TTQIPSSIED IFNDDRCINI TKQTPSFWIL ARALKEFVAK EGQGNLPVRG TIPDMIADSG KYIKLQNVYR EKAKKDAAAV GNHVAKLLQS IGQAPESISE KELKLLCSNS AFLRVVRCRS LAEEYGLDTI NKDEIISSMD NPDNEIVLYL MLRAVDRFHK QQGRYPGVSN YQVEEDIGKL KSCLTGFLQE YGLSVMVKDD YVHEFCRYGA AEPHTIAAFL GGAAAQEVIK IITKQFVIFN NTYIYSGMSQ TSATFQL | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NAE1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 10% glycerol and 200mM NaCl. |