missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NUDT9 Protein
A cDNA sequence encoding the NUDT9 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12908-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NUDT9 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MNTNVMSGSN GSKENSHNKA RTSPYPGSKV ERSQVPNEKV GWLVEWQDYK PVEYTAVSVL AGPRWADPQI SESNFSPKFN EKDGHVERKS KNGLYEIENG RPRNPAGRTG LVGRGLLGRW GPNHAADPII TRWKRDSSGN KIMHPVSGKH ILQFVAIKRK DCGEWAIPGG MVDPGEKISA TLKREFGEEA LNSLQKTSAE KREIEEKLHK LFSQDHLVIY KGYVDDPRNT DNAWMETEAV NYHDETGEIM DNLMLEAGDD AGKVKWVDIN DKLKLYASHS QFIKLVAEKR DAHWSEDSEA DCHAL | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NUDT9 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT and 0.1M NaCl. |