missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human OCT Protein
A cDNA sequence encoding the OCT was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12920-25mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
OCT Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
H-D-Phe-Cys-Phe-D-Trp-Lys-Thr-Cys-L-threoninol | |
Greater than 98.0% as determined by RP-HPLC. |
25 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The Octreotide was lyophilized from a concentrated (1 mg/ml) solution with no additives. |