Learn More
R&D Systems™ Recombinant Human Oncostatin M (OSM), Animal-Free Protein
Product Code. 18765935
Change view
Click to view available options
Quantity:
50 μg
500 μg
Unit Size:
500µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18765935 500 μg 500µg
18796065 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18765935 Supplier R&D Systems™ Supplier No. Qk0490500
 Restricted Item
Verfügbar ab: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
No significant difference between EC50 of reference and test lots
TRUSTED_SUSTAINABILITY
Specifications
Accession Number P13725.2
For Use With (Application) Bioactivity
Formulation Lyophilized from acetonitrile/TFA
Gene ID (Entrez) 5008
Molecular Weight (g/mol) M.W. (Observed): Monomeric Oncostatin M (OSM) protein only, M.W. (Theroretical): 22 kDa
Quantity 500 μg
Source E. coli-derived human Oncostatin M/OSM protein
Storage Requirements Store lyophilized protein between -20°C to -80°C until the date of expiry. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias MGC20461, oncostatin M, oncostatin-M
Conjugate Unconjugated
Reconstitution Resuspend in water at >100 μg/mL, prepare single use aliquots, add carrier protein if desired.
Product Line R&D Systems
Protein Oncostatin M/OSM