missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PFN4 Protein
A cDNA sequence encoding the PFN4 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13022-5ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
PFN4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSHLQSLLLD TLLGTKHVDS AALIKIQERS LCVASPGFNV TPSDVRTLVN GFAKNPLQAR REGLYFKGKD YRCVRADEYS LYAKNENTGV VVVKTHLYLL VATYTEGMYP SICVEATESL GDYLRKKGS | |
Greater than 95% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PFN4 Human solution containing 20mM Trsi HCL pH 8.0, 1mM DTT and 10% glycerol. |