missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPCS Protein
A cDNA sequence encoding the PPCS was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13116-5ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
PPCS Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAEMDPVAEF PQPPGAARWA EVMARFAARL GAQGRRVVLV TSGGTKVPLE ARPVRFLDNF SSGRRGATSA EAFLAAGYGV LFLYRARSAF PYAHRFPPQT WLSALRPSGP ALSGLLSLEA EENALPGFAE ALRSYQEAAA AGTFLAVEFT TLADYLHLLQ AAAQALNPLG PSAMFYLAAA VSDFYVPVSE MPEHKIQSSG GPLQITMKMV PKLLSPLVKD WAPKAFIISF KLETDPAIVI NRARKALEIY QHQVVVANIL ESRQSFVFIV TKDSETKLLL SEEEIEKGVE IEEKIVDNLQ SRHTAFIGDR N | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PPCS protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |