Learn More
enQuireBio™ Recombinant Human Protein-lysine 6-oxidase Protein
A DNA sequence encoding the Homo sapiens (Human) Protein-lysine 6-oxidase, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP8882-ec-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_002308.2 | |
Protein-lysine 6-oxidase Protein | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
4015 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Lox | |
Recombinant Protein | |
E. coli | |
PYKYSDDNPY YNYYDTYERP RPGGRYRPGY GTGYFQYGLP DLVADPYYIQ ASTYVQKMSM YNLRCAAEEN CLASTAYRAD VRDYDHRVLL RFPQRVKNQG TSDFLPSRPR YSWEWHSCHQ HYHSMDEFSH YDLLDANTQR RVAEGHKASF CLEDTSCDYG YHRRFACTAH TQGLSPGCYD TYGADIDCQW IDITDVKPGN YILKVSVNPS YLVPESDYTN NVVRCDIRYT GHHAYASGCT ISPY |