missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RARRES2 / Chemerin Protein
Recombinant Protein
Marke: enQuireBio™ QP5349-500ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_002880.1 | |
RARRES2 / Chemerin Protein | |
Research Use Only | |
RARRES2 | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
5919 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MELTEAQRRG LQVALEEFHK HPPVQWAFQE TSVESAVDTP FPAGIFVRLE FKLQQTSCRK RDWKKPECKV RPNGRKRKCL ACIKLGSEDK VLGRLVHCPI ETQVLREAEE HQETQCLRVQ RAGEDPHSFY FPGQFAFS | |
Greater than 95% as determined by SDS-PAGE. |