missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RHOG Protein
A cDNA sequence encoding the RHOG was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13318-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
RHOG Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMQS IKCVVVGDGA VGKTCLLICY TTNAFPKEYI PTVFDNYSAQ SAVDGRTVNL NLWDTAGQEE YDRLRTLSYP QTNVFVICFS IASPPSYENV RHKWHPEVCH HCPDVPILLV GTKKDLRAQP DTLRRLKEQG QAPITPQQGQ ALAKQIHAVR YLECSALQQD GVKEVFAEAV RAVLNPTPIK RGRSC | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RHOG solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 5mM DTT and 40% glycerol. |