missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100B Protein
A cDNA sequence encoding the S100B was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13402-20ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
6285 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MSELEKAMVA LIDVFHQYSG REGDKHKLKK SELKELINNE LSHFLEEIKE QEVVDKVMET LDNDGDGECD FQEFMAFVAM VTTACHEFFE HE |
S100B Protein | |
Research Use Only | |
S100B | |
Human | |
Untagged | |
S100B is supplied in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |