missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SCF / C-kit ligand Protein
Recombinant Protein
Marke: enQuireBio™ QP5225-500ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_000890.1 | |
SCF / C-kit ligand Protein | |
Research Use Only | |
KITLG | |
Recombinant Protein | |
E. coli | |
MEGICRNRVT NNVKDVTKLV ANLPKDYMIT LKYVPGMDVL PSHCWISEMV VQLSDSLTDL LDKFSNISEG LSNYSIIDKL VNIVDDLVEC VKENSSKDLK KSFKSPEPRL FTPEEFFRIF NRSIDAFKDF VVASETSDCV VSSTLSPEKD SRVSVTKPFM LPPVA | |
Greater than 95% as determined by SDS-PAGE. |
4254 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 15 ng/mL in the TF-1 cell proliferation assay. The bioactivity acceptance criteria is set at 6.6 x 10^4 units/mg. | |
Human | |
Untagged | |
10 mM acetic acid |