missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SLA2 Protein
A cDNA sequence encoding the SLA2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13517-5ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
SLA2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGSLPSR RKSLPSPSLS SSVQGQGPVT MEAERSKATA VALGSFPAGG PAELSLRLGE PLTIVSEDGD WWTVLSEVSG REYNIPSVHV AKVSHGWLYE GLSREKAEEL LLLPGNPGGA FLIRESQTRR GSYSLSVRLS RPASWDRIRH YRIHCLDNGW LYISPRLTFP SLQALVDHYS ELADDICCLL KEPCVLQRAG PLPGKDIPLP VTVQRTPLNW KELDSSLLFS EAATGEESLL SEGLRESLSF YISLNDEAVS LDDA | |
Greater than 80.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SLA2 protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 10% glycerol and 0.4M Urea. |