missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STX2 Protein
A cDNA sequence encoding the STX2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13634-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
STX2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMMRDRL PDLTACRKND DGDTVVVVEK DHFMDDFFHQ VEEIRNSIDK ITQYVEEVKK NHSIILSAPN PEGKIKEELE DLNKEIKKTA NKIRAKLKAI EQSFDQDESG NRTSVDLRIR RTQHSVLSRK FVEAMAEYNE AQTLFRERSK GRIQRQLEIT GRTTTDDELE EMLESGKPSI FTSDIISDSQ ITRQALNEIE SRHKDIMKLE TSIRELHEMF MDMAMFVETQ GEMINNIERN VMNATDYVEH AKEETKKAIK YQSKARRKK | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The STX2 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 50% glycerol. |