missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SUMO2 Protein
A cDNA sequence encoding the SUMO2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13649-50ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
6613 | |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MADEKPKEGV KTENNDHINL KVAGQDGSVV QFKIKRHTPL SKLMKAYCER QGLSMRQIRF RFDGQPINET DTPAQLEMED EDTIDVFQQQ TGGVY |
SUMO2 Protein | |
Research Use Only | |
SUMO2 | |
Human | |
Untagged | |
SUMO2 at 100microg/ml in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |