missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SYF2 Protein
A cDNA sequence encoding the SYF2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13655-5ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
SYF2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSDYEKVKL LEISAEDAER WERKKKRKNP DLGFSDYAAA QLRQYHRLTK QIKPDMETYE RLREKHGEEF FPTSNSLLHG THVPSTEEID RMVIDLEKQI EKRDKYSRRR PYNDDADIDY INERNAKFNK KAERFYGKYT AEIKQNLERG TAV | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SYF2 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 50% glycerol, 0.2M NaCl, 5mM DTT and 2mM EDTA. |