missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SYT11 Protein
A cDNA sequence encoding the SYT11 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13658-2ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
23208 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSWSCCHQQ AEKKQKNPPY KFIHMLKGIS IYPETLSNKK KIIKVRRDKD GPGREGGRRN LLVDAAEAGL LSRDKDPRGP SSGSCIDQLP IKMDYGEELR SPITSLTPGE SKTTSPSSPE EDVMLGSLTF SVDYNFPKKA LVVTIQEAHG LPVMDDQTQG SDPYIKMTIL PDKRHRVKTR VLRKTLDPVF DETFTFYGIP YSQLQDLVLH FLVLSFDRFS RDDVIGEVMV PLAGVDPSTG KVQLTRDIIK RNIQKCISRG ELQVSLSYQP VAQRMTVVVL KARHLPKMDI TGLSGNPYVK VNVYYGRKRI AKKKTHVKKC TLNPIFNESF IYDIPTDLLP DISIEFLVID FDRTTKNEVV GRLILGAHSV TASGAEHWRE VCESPRKPVA KWHSLSEY | |
Greater than 80% as determined by SDS-PAGE. |
SYT11 Protein | |
Research Use Only | |
SYT11 | |
Human | |
His | |
SYT11 protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 50% glycerol. |