missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TRAF1 Protein
A cDNA sequence encoding the TRAF1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13802-5ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
TRAF1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MDGTFLWKIT NVTRRCHESA CGRTVSLFSP AFYTAKYGYK LCLRLYLNGD GTGKRTHLSL FIVIMRGEYD ALLPWPFRNK VTFMLLDQNN REHAIDAFRP DLSSASFQRP QSETNVASGC PLFFPLSKLQ SPKHAYVKDD TMFLKCIVET ST | |
Greater than 95% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TRAF1 Human solution containing 20mM Tris-HCl pH 8.0, 0.1M NaCl, 1mM DTT and 20% glycerol. |