missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcriptional adapter 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Transcriptional adapter 3, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP8449-ec-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_001265199.1 | |
Transcriptional adapter 3 Protein | |
Research Use Only | |
TADA3 | |
Recombinant Protein | |
E. coli | |
MSELKDCPLQ FHDFKSVDHL KVCPRYTAVL ARSEDDGIGI EELDTLQLEL ETLLSSASRR LRVLEAETQI LTDWQDKKGD RRFLKLGRDH ELGAPPKHGK PKKQKLEGKA GHGPGPGPGR PKSKNLQPKI QEYEFTDDPI DVPRIPKNDA PNRFWASVEP YCADITSEEV RTLEELLKPP EDEAEHYKIP PLGKHYSQRW AQEDLLEEQK DGARAAAVAD KKKGLMGPLT ELDTKDVD | |
Greater than 90% as determined by SDS-PAGE. |
10474 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |