missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VAMP8 Protein
A cDNA sequence encoding the VAMP8 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13920-25ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
VAMP8 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEEASEGGGN DRVRNLQSEV EGVKNIMTQN VERILARGEN LEHLRNKTED LEATSEHFKT TSQKVARKFW WKNVKM | |
Greater than 85.0% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The VAMP8 protein solution (0.25 mg/ml) contains 20mM Tris pH 8.0, 0.1mM PMSF, 0.2M NaCl and 50% glycerol. |