missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VRK3 Protein
A cDNA sequence encoding the VRK3 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13947-10ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
VRK3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMISFCPD CGKSIQAAFK FCPYCGNSLP VEEHVGSQTF VNPHVSSFQG SKRGLNSSFE TSPKKVKWSS TVTSPRLSLF SDGDSSESED TLSSSERSKG SGSRPPTPKS SPQKTRKSPQ VTRGSPQKTS CSPQKTRQSP QTLKRSRVTT SLEALPTGTV LTDKSGRQWK LKSFQTRDNQ GILYEAAPTS TLTCDSGPQK QKFSLKLDAK DGRLFNEQNF FQRAAKPLQV NKWKKLYSTP LLAIPTCMGF GVHQDKYRFL VLPSLGRSLQ SALDVSPKHV LSERSVLQVA CRLLDALEFL HENEYVHGNV TAENIFVDPE DQSQVTLAGY GFAFRYCPSG KHVAYVEGSR SPHEGDLEFI SMDLHKGCGP SRRSDLQSLG YCMLKWLYGF LPWTNCLPNT EDIMKQKQKF VDKPGPFVGP CGHWIRPSET LQKYLKVVMA LTYEEKPPYA MLRNNLEALL QDLRVSPYDP IGLPMVP | |
Greater than 85% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The VRK3 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 40% glycerol. |