missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ZCCHC17 Protein
A cDNA sequence encoding the ZCCHC17 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13992-10ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
ZCCHC17 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMNSGRPE TMENLPALYT IFQGEVAMVT DYGAFIKIPG CRKQGLVHRT HMSSCRVDKP SEIVDVGDKV WVKLIGREMK NDRIKVSLSM KVVNQGTGKD LDPNNVIIEQ EERRRRSFQD YTGQKITLEA VLNTTCKKCG CKGHFAKDCF MQPGGTKYSL IPDEEEEKEE AKSAEFEKPD PTRNPSRKRK KEKKKKKHRD RKSSDSDSSD SESDTGKRAR HTSKDSKAAK KKKKKKKHKK KHKE | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ZCCHC17 protein solution (0.25 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 2mM DTT, 0.1mM PMSF and 20% glycerol. |