missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger protein 212 Protein
A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 212, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6905-ec-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_036388.2 | |
Zinc finger protein 212 Protein | |
Research Use Only | |
ZNF212 | |
Recombinant Protein | |
E. coli | |
MAESAPARHR RKRRSTPLTS STLPSQATEK SSYFQTTEIS LWTVVAAIQA VEKKMESQAA RLQSLEGRTG TAEKKLADCE KMAVEFGNQL EGKWAVLGTL LQEYGLLQRR LENVENLLRN RNFWILRLPP GSKGEAPKVS RSLENDGVCF TEQEWENLED WQKELYRNVM ESNYETLVSL KVLGQTEGEA ELGTEMLGDL EEEGPGGAHP AGGVMIKQEL QYTQEGPADL PGEFSCIAEE QAFLSPEQTE LWGGQGSSVL LETGPGDSTL EEPVGSRVPS SSRTVGCPKQ KSHRQVQLDQ ECGQGLKLKK DTSRPYECSE CEITFRYKQQ LATHLRSHSG WGSCTPEEPE ESLRPRPRLK PQTKKAKLHQ CDVCLRSFSC KVSLVTHQRC HLQEGPSAGQ HVQERFSPNS LVALPGHIPW RKSRSSLICG YCGKSFSHPS DLVRHQRIHT GERPYSCTEC EKSFVQKQHL LQHQKIHQRE RGGLALEPGR PNGLL | |
Greater than 90% as determined by SDS-PAGE. |
7988 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |