missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque Erythropoietin / EPO Protein
A DNA sequence encoding the Macaca mulatta (Rhesus macaque) Erythropoietin / EPO, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10075-ma-20ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_001036201.1 | |
Erythropoietin / EPO Protein | |
Research Use Only | |
EPO | |
Recombinant Protein | |
Mammal | |
APPRLVCDSR VLERYLLEAK EAENVTMGCS ESCSLNENIT VPDTKVNFYA WKRIEVGQQA VEVWQGLALL SEAVLRGQAV LANSSQPFEP LQLHMDKAIS GLRSITTLLR ALGAQEAISL PDAASAAPLR TITADTFCKL FRVYSNFLRG KLKLYTGEAC RRGDR | |
Greater than 90% as determined by SDS-PAGE. |
719294 | |
20 μg | |
Not determined. | |
Full Length | |
Macaque | |
His | |
Tris-based buffer, 50% glycerol |