Learn More
enQuireBio™ Recombinant Mouse 60S acidic ribosomal protein P0 Protein
A DNA sequence encoding the Mus musculus (Mouse) 60S acidic ribosomal protein P0, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6624-ec-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_031501.1 | |
60S acidic ribosomal protein P0 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
11837 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
RPLP0 | |
Recombinant Protein | |
E. coli | |
MPREDRATWK SNYFLKIIQL LDDYPKCFIV GADNVGSKQM QQIRMSLRGK AVVLMGKNTM MRKAIRGHLE NNPALEKLLP HIRGNVGFVF TKEDLTEIRD MLLANKVPAA ARAGAIAPCE VTVPAQNTGL GPEKTSFFQA LGITTKISRG TIEILSDVQL IKTGDKVGAS EATLLNMLNI SPFSFGLIIQ QVFDNGSIYN PEVLDITEQA LHSRFLEGVR NVASVCLQIG YPTVASVPHS IINGYKRVLA LSVETEYTFP LTEKVKAFLA DPSAFAAAAP AAAATTAAPA AAAAPAKAEA KEESEESDED MGFGLFD |