missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse ACY1 Protein
A cDNA sequence encoding the ACY1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10957-10ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
109652 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEFMTTKD PESEHPSVTL FRQYLRICTV QPNPDYGGAI TFLEERARQL GLSCQKIEVV PGFVITVLTW PGTNPSLPSI LLNSHTDVVP VFKEHWHHDP FEAFKDSEGY IYARGSQDMK SVSIQYLEAV RRLKSEGHRF PRTIHMTFVP DEEVGGHKGM ELFVKRPEFQ ALRAGFALDE GLANPTDAFT VFYSERSPWW VRVTSTGKPG HASRFIEDTA AEKLHKVISS ILAFREKERQ RLQANPHLKE GAVTSVNLTK LEGGVAYNVV PATMSASFDF RVAPDVDMKA FEKQLQRWCQ EAGEGVTFEF AQKFTEPRMT PTDDSDPWWA AFSGACKAMN LTLEPEIFPA ATDSRYIRAV GIPALGFSPM NRTPVLLHDH NERLHEDIFL RGVDIYTGLL SALASVPTLP GES | |
Greater than 90.0% as determined by SDS-PAGE. |
ACY1 Protein | |
Research Use Only | |
Acy1 | |
Mouse | |
His | |
ACY1 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4) and 10% glycerol. |