missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse CD155/PVR His-tag Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke: R&D Systems™ 6909-CD-050
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Measured by its binding ability in a functional ELISA. When Recombinant Mouse CD155/PVR is coated at 5 μg/mL (100 μL/well), the concentration of Recombinant Mouse CD96 (Catalog # 5690-CD ) that produces 50% optimal binding response is found to be approximately 0.015-0.075 μg/mL.Spezifikation
NP_081790 | |
Lyophilized from a 0.2 μm filtered solution in PBS. | |
M.W. (Observed): 63-80 kDa, reducing conditions; M.W. (theoretical): 35.6 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
Pvr | |
Recombinant | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Bioactivity | |
5817 | |
CD155/PVR | |
Mouse myeloma cell line, NS0-derived mouse CD155/PVR protein Met1-Leu348, with a C-terminal 6-His tag | |
<0.01 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
Reconstitute at 100 μg/mL in PBS. | |
CD155/PVR |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur