missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse FLT3L / Flt3 ligand Protein
Recombinant Protein
Marke: enQuireBio™ QP5405-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_038548.3 | |
FLT3L / Flt3 ligand Protein | |
Research Use Only | |
Flt3l | |
Recombinant Protein | |
E. coli | |
MTPDCYFSHS PISSNFKVKF RELTDHLLKD YPVTVAVNLQ DEKHCKALWS LFLAQRWIEQ LKTVAGSKMQ TLLEDVNTEI HFVTSCTFQP LPECLRFVQT NISHLLKDTC TQLLALKPCI GKACQNFSRC LEVQCQPDSS TLLPPRSPIA LEATELPEPR PRQ | |
Greater than 95% as determined by SDS-PAGE. |
14256 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 10 ng/mL in the OCI-AML5 cell proliferation assay. The bioactivity acceptance criteria is set at 1 x 10^5 units/mg. | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |