missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL-12AB Protein
Recombinant Protein
Marke: enQuireBio™ QP5204-2ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_001290173.1 | |
IL-12AB Protein | |
Research Use Only | |
IL12 | |
Recombinant Protein | |
Human (HEK293) | |
p35:MRVIPV SGPARCLSQS RNLLKTTDDM VKTAREKLKH YSCTAEDIDH EDITRDQTST LKTCLPLELH KNESCLATRE TSSTTRGSCL PPQKTSLMMT LCLGSIYEDL KMYQTEFQAI NAALQNHNHQ QIILDKGMLV AIDELMQSLN HNGETLRQKP PVGEADPYRV KMKLCILLHA FSTRVVTINR VMGYLSSA p40:MWELEKDV YVVEVDWTPD APGETVNLTC DTPEEDDITW TSDQRHGVIG SGKTLTITVK EFLDAGQYTC HKGGETLSHS HLLLHKKENG IWSTEILKNF KNKTFLKCEA PNYSGRFTCS WLVQRNMDLK FNIKSSSSSP DSRAVTCGMA SLSAEKVTLD QRDYEKYSVS CQEDVTCPTA EETLPIELAL EARQQNKYEN YSTSFFIRDI IKPDPPKNLQ MKPLKNSQVE VSWEYPDSWS TPHSYFSLKF FVRIQRKKEK MKETEEGCNQ KGAFLVEKTS TEVQCKGGNV CVQAQDRYYN SSCSKWACVP CRVRS | |
Greater than 95% as determined by SDS-PAGE. |
16160 | |
2 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 20 ng/mL in the Mouse primary T cell induced expression of CD25 assay. The bioactivity acceptance criteria is set at 5 x 10^4 units/mg. | |
Mouse | |
Untagged | |
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5 |