missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL-3 / Interleukin-3 Protein
Recombinant Protein
Marke: enQuireBio™ QP4199-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_034686.2 | |
IL-3 / Interleukin-3 Protein | |
Research Use Only | |
IL3 | |
Recombinant Protein | |
E. coli | |
MDTHRLTRTL NCSSIVKEII GKLPEPELKT DDEGPSLRNK SFRRVNLSKF VESQGEVDPE DRYVIKSNLQ KLNCCLPTSA NDSALPGVFI RDLDDFRKKL RFYMVHLNDL ETVLTSRPPQ PASGSVSPNR GTVEC | |
Greater than 95% as determined by SDS-PAGE. |
16187 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 250 pg/mL in the NFS-60 cell proliferation assay. The bioactivity acceptance criteria is set at 4 x 10^6 units/mg. | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |