missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL13 / ALRH Protein
Recombinant Protein
Marke: enQuireBio™ QP4168-500ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_032381.1 | |
IL13 / ALRH Protein | |
Research Use Only | |
IL13 | |
Recombinant Protein | |
E. coli | |
MPVPRSVSLP LTLKELIEEL SNITQDQTPL CNGSMVWSVD LAAGGFCVAL DSLTNISNCN AIYRTQRILH GLCNRKAPTT VSSLPDTKIE VAHFITKLLS YTKQLFRHGP F | |
Greater than 95% as determined by SDS-PAGE. |
16163 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 10 ng/mL in the TF-1 cell proliferation assay. The bioactivity acceptance criteria is set at 1 x 10^5 units/mg. | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |