missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL33 / Interleukin-33 / NF-HEV Protein
Recombinant Protein
Marke: enQuireBio™ QP5444-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_001158196.1 | |
IL33 / Interleukin-33 / NF-HEV Protein | |
Research Use Only | |
IL33 | |
Recombinant Protein | |
E. coli | |
MSIQGTSLLT QSPASLSTYN DQSVSFVLEN GCYVINVDDS GKDQEQDQVL LRYYESPCPA SQSGDGVDGK KLMVNMSPIK DTDIWLHAND KDYSVELQRG DVSPPEQAFF VLHKKSSDFV SFECKNLPGT YIGVKDNQLA LVEEKDESCN NIMFKLSKI | |
Greater than 90 as determined by SDS-PAGE. |
77125 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 2 ng/mL in the D10S cell proliferation assay. The bioactivity acceptance criteria is set at 5 x 10^5 units/mg. | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |