missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MYL9 Protein
A cDNA sequence encoding the MYL9 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12774-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Specifications
MYL9 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSSKRA KAKTTKKRPQ RATSNVFAMF DQSQIQEFKE AFNMIDQNRD GFIDKEDLHD MLASLGKNPT DEYLEGMMNE APGPINFTMF LTMFGEKLNG TDPEDVIRNA FACFDEEASG FIHEDHLREL LTTMGDRFTD EEVDEMYREA PIDKKGNFNY VEFTRILKHG AKDKDD | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
MYL9 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |