missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse NPM1 / Nucleophosmin Protein
A DNA sequence encoding the Mus musculus (Mouse) NPM1 / Nucleophosmin, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP7731-ec-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_032748.1 | |
NPM1 / Nucleophosmin Protein | |
Research Use Only | |
Npm1 | |
Recombinant Protein | |
E. coli | |
MEDSMDMDMS PLRPQNYLFG CELKADKDYH FKVDNDENEH QLSLRTVSLG AGAKDELHIV EAEAMNYEGS PIKVTLATLK MSVQPTVSLG GFEITPPVVL RLKCGSGPVH ISGQHLVAVE EDAESEDEDE EDVKLLGMSG KRSAPGGGNK VPQKKVKLDE DDEDDDEDDE DDEDDDDDDF DEEETEEKVP VKKSVRDTPA KNAQKSNQNG KDLKPSTPRS KGQESFKKQE KTPKTPKGPS SVEDIKAKMQ ASIEKGGSLP KVEAKFINYV KNCFRMTDQE AIQDLWQWRK SL | |
Greater than 90% as determined by SDS-PAGE. |
18148 | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |