missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse PIN1 Protein
A cDNA sequence encoding the PIN1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10819-10ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
23988 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 1,200 nmol/min/mg, and is defined as the amount of enzyme that cleaves 1nmole of suc-AAFP-PNA per minute at 37C in Tris-HCl pH 8.0 using chymotrypsin. | |
Mouse | |
His | |
PIN1 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4) and 10% glycerol. |
PIN1 Protein | |
Research Use Only | |
Pin1 | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMADEEKL PPGWEKRMSR SSGRVYYFNH ITNASQWERP SGGSTVGGSS KNGQGEPAKV RCSHLLVKHS QSRRPSSWRQ EKITRSKEEA LELINGYIQK IKSGEEDFES LASQFSDCSS AKARGDLGPF SRGQMQKPFE DASFALRTGE MSGPVFTDSG IHIILRTE | |
Greater than 95.0% as determined by SDS-PAGE. |