missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse S100A10 Protein
A cDNA sequence encoding the S100A10 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13387-2ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
20194 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPSQMEH AMETMMLTFH RFAGDKDHLT KEDLRVLMER EFPGFLENQK DPLAVDKIMK DLDQCRDGKV GFQSFLSLVA GLTIACNDYF VVNMKQKGKK | |
Greater than 90.0% as determined by SDS-PAGE. |
S100A10 Protein | |
Research Use Only | |
S100a10 | |
Mouse | |
His | |
S100A10 protein solution (1 mg/ml) containing phosphate buffered saline (pH 7.4) and 10% glycerol. |