missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Serpin H1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Serpin H1, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6677-ec-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_001104513.1 | |
Serpin H1 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
12406 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Serpinh1 | |
Recombinant Protein | |
E. coli | |
AEVKKPLEAA APGTAEKLSS KATTLAERST GLAFSLYQAM AKDQAVENIL LSPLVVASSL GLVSLGGKAT TASQAKAVLS AEKLRDEEVH TGLGELLRSL SNSTARNVTW KLGSRLYGPS SVSFADDFVR SSKQHYNCEH SKINFRDKRS ALQSINEWAS QTTDGKLPEV TKDVERTDGA LLVNAMFFKP HWDEKFHHKM VDNRGFMVTR SYTVGVTMMH RTGLYNYYDD EKEKLQMVEM PLAHKLSSLI ILMPHHVEPL ERLEKLLTKE QLKAWMGKMQ KKAVAISLPK GVVEVTHDLQ KHLAGLGLTE AIDKNKADLS RMSGKKDLYL ASVFHATAFE WDTEGNPFDQ DIYGREELRS PKLFYADHPF IFLVRDNQSG SLLFIGRLVR PKGDKMRDEL |