missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SERPINA6 Protein
A cDNA sequence encoding the SERPINA6 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13459-10ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
SERPINA6 Protein | |
Research Use Only | |
Recombinant Protein | |
Human (HEK293) | |
VTDEDSSSHR DLAPTNVDFA FNLYKRLVAL NSDKNTLISP VSISMALAML SLSTRGSTQY LENLGFNMSK MSEAEIHQGF QYLNSLLQQS DTGLEMNMGN VMFLLQNLKL KDSFLADTKH YYESEALTIP SKDWTKAGEQ INNHVKNKTQ GKIEHVVSDL DSSATLILIN YIFLKGIWKL PFSPENTREE DFYVNETSTV KVPMMVQSGN ISYFRDSAIP CQMVQMNYVG NGTTFIILPD QGQMDTVVAA LNRDTIDRWG KLMIPRQMNL YIPKFSMSDT YDLQDVLADV GIKDLFTNQS DFADTTKDTP LTLTVLHKAM LQLDEGNVLP AATNGPPVHL PSESFTLKYN RPFIFLAFDK YTWSSLMMSQ VMNPA HHHHH HHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
SERPINA6 was filtered (0.4microm) and lyophilized in phosphate buffered saline pH 8.0. |