Learn More
enQuireBio™ Recombinant Mouse Serum amyloid A-2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Serum amyloid A-2 protein, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6649-ec-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_035444.1 | |
Serum amyloid A-2 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
20209 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Saa2 | |
Recombinant Protein | |
E. coli | |
GFFSFIGEAF QGAGDMWRAY TDMKEAGWKD GDKYFHARGN YDAAQRGPGG VWAAEKISDA RESFQEFFGR GHEDTMADQE ANRHGRSGKD PNYYRPPGLP AKY |