missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Borrelia OspC Protein
A cDNA sequence encoding the Borrelia OspC was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11190-20ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Borrelia OspC Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Borrelia OspC (1.74 mg/1ml) is supplied in 16mM HEPES buffer pH 7.0, 300mM NaCl and 20% glycerol. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Greater than 80.0% as determined by SDS-PAGE. |