missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Hexarelin Protein
A cDNA sequence encoding the Hexarelin was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12225-20mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Hexarelin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The Hexarelin peptide was lyophilized with no additives. |
20 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |