missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other lldD Protein
A cDNA sequence encoding the lldD was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12578-10ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
948121 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMIISAA SDYRAAAQRI LPPFLFHYMD GGAYSEYTLR RNVEDLSEVA LRQRILKNMS DLSLETTLFN EKLSMPVALA PVGLCGMYAR RGEVQAAKAA DAHGIPFTLS TVSVCPIEEV APAIKRPMWF QLYVLRDRGF MRNALERAKA AGCSTLVFTV DMPTPGARYR DAHSGMSGPN AAMRRYLQAV THPQWAWDVG LNGRPHDLGN ISAYLGKPTG LEDYIGWLGN NFDPSISWKD LEWIRDFWDG PMVIKGILDP EDARDAVRFG ADGIVVSNHG GRQLDGVLSS ARALPAIADA VKGDIAILAD SGIRNGLDVV RMIALGADTV LLGRAFLYAL ATAGQAGVAN LLNLIEKEMK VAMTLTGAKS ISEITQDSLV QGLGKELPAA LAPMAKGNAA | |
Greater than 90% as determined by SDS-PAGE. |
lldD Protein | |
Research Use Only | |
lldD | |
Other | |
His | |
The lldD solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 1mM DTT. |