missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Ovine Leptin Protein
A cDNA sequence encoding the Leptin was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10750-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Leptin Protein | |
Research Use Only | |
Biological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. | |
Ovine | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3. |
100 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
VPIRKVQDDT KTLIKTIVTR INDISHTQSV SSKQRVTGLD FIPGLHPLLS LSKMDQTLAI YQQILASLPS RNVIQISNDL ENLRDLLHLL AASKSCPLPQ VRALESLESL GVVLEASLYS TEVVALSRLQ GSLQDMLRQL DLSPGC | |
Greater than 95.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |