Learn More
enQuireBio™ Recombinant P. aeruginosa UDP-3-O-acyl-N-acetylglucosamine deacetylase Protein
A DNA sequence encoding the Pseudomonas aeruginosa (strain ATCC 15692 / DSM 22644 / CIP 104116 / JCM 14847 / LMG 12228 / 1C / PRS 101 / PAO1) UDP-3-O-acyl-N-acetylglucosamine deacetylase, was expressed in the hosts and tags indicated. P
Marke: enQuireBio™ QP7215-ec-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_253096.1 | |
P. aeruginosa UDP-3-O-acyl-N-acetylglucosamine deacetylase Protein | |
Research Use Only | |
lpxC | |
Recombinant Protein | |
E. coli | |
MIKQRTLKNI IRATGVGLHS GEKVYLTLKP APVDTGIVFC RTDLDPVVEI PARAENVGET TMSTTLVKGD VKVDTVEHLL SAMAGLGIDN AYVELSASEV PIMDGSAGPF VFLIQSAGLQ EQEAAKKFIR IKREVSVEEG DKRAVFVPFD GFKVSFEIDF DHPVFRGRTQ QASVDFSSTS FVKEVSRART FGFMRDIEYL RSQNLALGGS VENAIVVDEN RVLNEDGLRY EDEFVKHKIL DAIGDLYLLG NSLIGEFRGF KSGHALNNQL LRTLIADKDA WEVVTFEDAR TAPISYMRPA AAV | |
Greater than 80% as determined by SDS-PAGE. |
881292 | |
100 μg | |
Not determined. | |
Full Length | |
P. aeruginosa | |
His-SUMO | |
Tris-based buffer, 50% glycerol |