missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat BD 1 Protein
A cDNA sequence encoding the BD 1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10531-20ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
83687 | |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Measured by its ability to chemoattract CD34+ dendritic cells using a concentration range of 0.1-1.0 ug/ml. | |
Rat | |
Untagged | |
BD-1 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |
BD 1 Protein | |
Research Use Only | |
Defb1 | |
Recombinant Protein | |
E. coli | |
DQYRCLQNGG FCLRSSCPSH TKLQGTCKPD KPNCCRS | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |