missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CALB1 Protein
A cDNA sequence encoding the CALB1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11257-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
83839 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MAESHLQSSL ITASQFFEIW LHFDADGSGY LEGKELQNLI QELLQARKKA GLELSPEMKT FVDQYGQRDD GKIGIVELAH VLPTEENFLL LFRCQQLKSC EEFMKTWRKY DTDHSGFIET EELKNFLKDL LEKANKTVDD TKLAEYTDLM LKLFDSNNDG KLELTEMARL LPVQENFLLK FQGIKMCGKE FNKAFELYDQ DGNGYIDENE LDALLKDLCE KNKQELDINN ISTYKKNIMA LSDGGKLYRT DLALILSAGD N | |
Greater than 90% as determined by SDS-PAGE. |
CALB1 Protein | |
Research Use Only | |
Calb1 | |
Rat | |
Untagged | |
The protein was lyophilized from a concentrated solution (1 mg/ml) containing 50mM NaHCO3. |