missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat KGF 2 Protein
A cDNA sequence encoding the KGF 2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10736-5ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
KGF 2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
QALGQDMVSP EATNSSSSSS SSSSSSSFSS PSSAGRHVRS YNHLQGDVRW RKLFSFTKYF LKIEKNGKVS GTKKENCPYS ILEITSVEIG VVAVKAINSN YYLAMNKKGK LYGSKEFNND CKLKERIEEN GYNTYASFNW QHNGRQMYVA LNGKGAPRRG QKTRRKNTSA HFLPMVVHS | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Rat | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4 containing 5% trehalose. |