missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat TFF3 Protein
A cDNA sequence encoding the TFF3 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13729-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
25563 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MQEFVGLSPS QCMVPANVRV DCGYPTVTSE QCNNRGCCFD SSIPNVPWCF KPLQETECT F DYKDDDDK | |
Greater than 95.0% as determined by SDS-PAGE. |
TFF3 Protein | |
Research Use Only | |
Tff3 | |
Rat | |
Untagged | |
TFF3 protein filtered (0.4microm) and lyophilized in 0.5 mg/ml in 20mM TRIS and 50mM NaCl, pH 7.5. |